Learn More
Biomatik Corporation Human Podocalyxin, Pcx Elisa Kit
Supplier:  Biomatik Corporation EKC35061
Encompass_Preferred
Human Podocalyxin, Pcx Elisa Kit for detecting Podocalyxin, Pcx in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.54-400ng/ml. Sensitivity: 0.54ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O00592; PODXL. Target Synonyms: GCTM-2 antigen; Gp2; Gp200; MGC138240; PC; PCLP; PCLP-1; PCLP1; Pcx; Podocalyxin; Podocalyxin like; Podocalyxin like protein; Podocalyxin-like protein 1; Podxl; PODXL_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7521