Learn More
Biomatik Corporation Human procollagen type III ,HPCⅢ ELISA kit
Supplier:  Biomatik Corporation EKC35113
Encompass_Preferred
Sample Type: serum. Species Reactivity: Homo sapiens (Human). Detection Range: 25 ng/mL-300 ng/mL. Sensitivity: 25 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: procollagen type 3 ,HPC3|procollagen type III ,HPCⅢ,procollagen type III ,HPCIII. Restriction: For Research Use Only.
Catalog No. 50-228-7523