Learn More
Biomatik Corporation Human Proteoglycan 4 (Prg4) Elisa Kit
Supplier:  Biomatik Corporation EKC35277
Encompass_Preferred
Human Proteoglycan 4 (Prg4) Elisa Kit for detecting Proteoglycan 4(Prg4) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92954; PRG4. Target Synonyms: PRG4; MSF; SZP; Proteoglycan 4; Lubricin; Megakaryocyte-stimulating factor; Superficial zone proteoglycan) [Cleaved into: Proteoglycan 4 C-terminal part]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7531