Learn More
Biomatik Corporation Human Tgf-Beta-Inducible Early Response Gene-1, Tieg1 Elisa Kit
Supplier:  Biomatik Corporation EKC35752
Encompass_Preferred
Human Tgf-Beta-Inducible Early Response Gene-1, Tieg1 Elisa Kit for detecting Kruppel-Like Factor 10 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 34.38-2200pg/ml. Sensitivity: 8.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13118; KLF10. Target Synonyms: Early growth response; alpha; Early growth response; alpha like; EGR A; EGR alpha; EGR (A); EGR-alpha; EGRA; Egral; EGRalpha; Gdnfif; KLF 10; KLF10; KLF10_HUMAN; Krueppel like factor 10; Krueppel like factor10; Krueppel-like factor 10; Kruppel like factor 10; mGIF; TGF-beta- inducible early gene; TGFB inducible early growth response 1; TGFB inducible early growth response; TGFB inducible early growth response protein 1; TGFB-inducible early growth response protein 1; TIEG 1; TIEG; TIEG-1; TIEG1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7550