Learn More
Biomatik Corporation Mouse anti-nucleosome antibody (IgG) ELISA Kit
Supplier:  Biomatik Corporation EKC36341
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 U/mL-20 U/mL. Sensitivity: 0.078 U/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7572