Learn More
Biomatik Corporation Mouse Basic Fibroblast Growth Factor, Bfgf Elisa Kit
Supplier:  Biomatik Corporation EKC36388
Encompass_Preferred
Mouse Basic Fibroblast Growth Factor, Bfgf Elisa Kit for detecting Fibroblast Growth Factor 2 (Basic) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P15655; Fgf2. Target Synonyms: Fgf2; Fgf-2Fibroblast growth factor 2; FGF-2; Basic fibroblast growth factor; bFGF; Heparin-binding growth factor 2; HBGF-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7575