Learn More
Biomatik Corporation Mouse Eosinophil Peroxidase (EPO) ELISA Kit
Supplier:  Biomatik Corporation EKC36802
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P49290. Target Synonyms: EPO, EPP, EPX-PEN. Restriction: For Research Use Only.
Catalog No. 50-228-7590