Learn More
Biomatik Corporation Mouse Heat Shock Protein 70,Hsp-70 ELISA Kit
Supplier:  Biomatik Corporation EKC37024
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q61696. Target Synonyms: DAQB-147D11.1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, HSP70-1, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B, dnaK-type molecular chaperone HSP70-1|heat shock 70kD protein 1A|heat shock-induced. Restriction: For Research Use Only.
Catalog No. 50-228-7596