Learn More
Biomatik Corporation Mouse intercellular adhesion molecule 1,ICAM-1 ELISA KIT
Supplier:  Biomatik Corporation EKC37130
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 78 ng/mL-5000 ng/mL. Sensitivity: 19.5 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P13597. Target Synonyms: BB2, CD54, P3.58, cell surface glycoprotein P3.58|human rhinovirus receptor|intercellular adhesion molecule 1 (CD54), human rhinovirus receptor. Restriction: For Research Use Only.
Catalog No. 50-228-7601