Learn More
Biomatik Corporation Mouse Interleukin 22 (Il-22) Elisa Kit
Supplier:  Biomatik Corporation EKC37159
Encompass_Preferred
Mouse Interleukin 22 (Il-22) Elisa Kit for detecting Interleukin 22 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9JJY9; Il22. Target Synonyms: Il22; Il22a; Iltif; IltifaInterleukin-22; IL-22; IL-10-related T-cell-derived-inducible factor; IL-TIF; IL-TIF alpha; Interleukin-22a; IL-22a. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7605