Learn More
Biomatik Corporation Mouse Lipoprotein-Associated Phospholipase A2, Lp-Pla2 Elisa Kit
Supplier:  Biomatik Corporation EKC37296
Encompass_Preferred
Mouse Lipoprotein-Associated Phospholipase A2, Lp-Pla2 Elisa Kit for detecting Lipoprotein-Associated Phospholipase A2, Lp-Pla2 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q60963; Pla2g7. Target Synonyms: Pla2g7; Pafah; Platelet-activating factor acetylhydrolase; PAF acetylhydrolase; EC 3.1.1.47; 1-alkyl-2-acetylglycerophosphocholine esterase; 2-acetyl-1-alkylglycerophosphocholine esterase; LDL-associated phospholipase A2; LDL-PLA (2); PAF 2-acylhydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7609