Learn More
Biomatik Corporation Mouse Orexin A Elisa Kit
Supplier:  Biomatik Corporation EKC37520
Encompass_Preferred
Mouse Orexin A Elisa Kit for detecting Orexin A in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O55241; Hcrt. Target Synonyms: Hcrt; Ox; Ppox; Orexin; Hypocretin; Hcrt) [Cleaved into: Orexin-A; Hypocretin-1; Hcrt1); Orexin-B; Hypocretin-2; Hcrt2)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7616