Learn More
Biomatik Corporation Mouse Α-Melanocyte Stimulating Hormone (Α-Msh) Elisa Kit
Supplier:  Biomatik Corporation EKC38068
Encompass_Preferred
Mouse Α-Melanocyte Stimulating Hormone (Α-Msh) Elisa Kit for detecting Α-Melanocyte Stimulating Hormone(Α-Msh) in serum, plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800pg/ml. Sensitivity: 3.12pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: alpha-melanocyte stimulating hormone; alpha-MSH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7627