Learn More
Biomatik Corporation Rat hypoxia-inducible factor 1α,HIF-1α ELISA Kit
Supplier:  Biomatik Corporation EKC39208
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 3.12 pg/mL-200 pg/mL. Sensitivity: 0.78 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O35800. Target Synonyms: HIF-1alpha, HIF1, HIF1-ALPHA, MOP1, PASD8, bHLHe78, ARNT interacting protein|hypoxia-inducible factor 1 alpha isoform I.3|hypoxia-inducible factor 1, alpha subunit|hypoxia-inducible factor 1, alpha|hypoxia-inducible factor 1α(HIF-1α)|hypoxia-inducible factor 1 alpha(HIF-1 alpha). Restriction: For Research Use Only.
Catalog No. 50-228-7664