Learn More
Biomatik Corporation Rat neuropeptide Y (NPY) ELISA kit
Supplier:  Biomatik Corporation EKC39480
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.56 pg/mL-100 pg/mL. Sensitivity: 0.39 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07808. Target Synonyms: PYY4, OTTHUMP00000201946|OTTHUMP00000201947. Restriction: For Research Use Only.
Catalog No. 50-228-7672