Learn More
Biomatik Corporation Dog Interleukin 18, Il-18 Elisa Kit
Supplier:  Biomatik Corporation EKC40020
Encompass_Preferred
Dog Interleukin 18, Il-18 Elisa Kit for detecting Interleukin 18 (Interferon-Gamma-Inducing Factor) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 12.5-800pg/ml. Sensitivity: 3.12pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9XSR0; IL18. Target Synonyms: IL18; IGIF; Interleukin-18; IL-18; Interferon gamma-inducing factor; IFN-gamma-inducing factor; Interleukin-1 gamma; IL-1 gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7688