Learn More
Biomatik Corporation Pig Interleukin 18,IL-18 ELISA Kit
Supplier:  Biomatik Corporation EKC40215
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O19073. Target Synonyms: IGIF, IL-18, IL-1g, IL1F4, MGC12320, IL-1 gamma|interferon-gamma-inducing factor|interleukin 18|interleukin-1 gamma|interleukin-18. Restriction: For Research Use Only.
Catalog No. 50-228-7714