Learn More
Biomatik Corporation Rat Interleukin 13, Il-13 Elisa Kit
Supplier:  Biomatik Corporation EKC40335
Encompass_Preferred
Rat Interleukin 13, Il-13 Elisa Kit for detecting Interleukin 13 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P42203; Il13. Target Synonyms: Il13; Il-13; Interleukin-13; IL-13; T-cell activation protein P600. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7720