Learn More
Biomatik Corporation Mouse Collagen Type Ⅳ,Col Ⅳ ELISA KIT
Supplier:  Biomatik Corporation EKC40536
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.78 ng/mL-50 ng/mL. Sensitivity: 0.195 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Collagen Type 4,Col 4,_x000D_,Collagen Type IV,Col IV. Restriction: For Research Use Only.
Catalog No. 50-228-7748