Learn More
Biomatik Corporation Mouse regulated on activation in normal T-cell expressed and secreted,RANTES ELISA kit
Supplier:  Biomatik Corporation EKC40593
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P30882. Target Synonyms: D17S136E, MGC17164, RANTES, SCYA5, SISd, TCP228, OTTHUMP00000197106|SIS-delta|T-cell specific protein p288|beta-chemokine RANTES|regulated upon activation, normally T-expressed, and presumably secre. Restriction: For Research Use Only.
Catalog No. 50-228-7759