Learn More
Biomatik Corporation Human Apoptosis Regulator Bax Elisa Kit
Supplier:  Biomatik Corporation EKC40612
Encompass_Preferred
Human Apoptosis Regulator Bax Elisa Kit for detecting Bcl2-Associated X Protein in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.312ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q07812; BAX. Target Synonyms: Apoptosis regulator BAX; BAX; Bax-protein; BAX_HUMAN; BAXA; Baxdelta2G9; Baxdelta2G9omega; Baxdelta2omega; Bcl-2-like protein 4; BCL2 associated X protein; BCL2 associated X protein omega; BCL2 associated X protein transcript variant delta2; Bcl2-L-4; BCL2L4; membrane isoform alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7763