Learn More
Biomatik Corporation Human Integrin Β2, Itg Β2/Cd11+Cd18 Elisa Kit
Supplier:  Biomatik Corporation EKC40724
Encompass_Preferred
Human Integrin Β2, Itg Β2/Cd11+Cd18 Elisa Kit for detecting Integrin, Beta 2 (Complement Component 3 Receptor 3 And 4 Subunit) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05107; ITGB2. Target Synonyms: ITGB2; CD18; MFI7; Integrin beta-2; Cell surface adhesion glycoproteins LFA-1/CR3/p150; 95 subunit beta; Complement receptor C3 subunit beta; CD antigen CD18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7774