Learn More
Biomatik Corporation Human Epidermal Growth Factor Receptor 2 (Sp185/Her2) Elisa Kit
Supplier:  Biomatik Corporation EKC40773
Encompass_Preferred
Human Epidermal Growth Factor Receptor 2 (Sp185/Her2) Elisa Kit for detecting V-Erb-B2 Erythroblastic Leukemia Viral Oncogene Homolog 2, Neuro/Glioblastoma Derived Oncogene Homolog (Avian) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.04ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04626; ERBB2. Target Synonyms: Verb b2 erythroblastic leukemia viral oncogene homolog 2; neuro/glioblastoma derived oncogene homolog; C erb B2/neu protein; CD340; CD340 antigen; Cerb B2/neu protein; CerbB2; Erb b2 receptor tyrosine kinase 2; ErbB-2 proto-oncogene; ERBB2; ERBB2_HUMAN; HER 2; HER 2/NEU; HER2; Herstatin; Human epidermal growth factor receptor 2; Metastatic lymph node gene 19 protein; MLN 19; MLN19; NEU; NEU proto oncogene). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7783