Learn More
Biomatik Corporation Human Vascular Endothelial Cell Growth Factor Receptor 1 (Vegfr-1/Flt1) Elisa Kit
Supplier:  Biomatik Corporation EKC40822
Encompass_Preferred
Human Vascular Endothelial Cell Growth Factor Receptor 1 (Vegfr-1/Flt1) Elisa Kit for detecting Fms-Related Tyrosine Kinase 1 (Vascular Endothelial Growth Factor/Vascular Permeability Factor Receptor) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P17948; FLT1. Target Synonyms: EC 2.7.10.1; FLT 1; FLT; Flt-1; FLT1; Fms like tyrosine kinase 1; Fms related tyrosine kinase 1; Fms related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); Fms related tyrosine kinase 1 vascular endothelial growth factor/vascular permeability factor receptor; Fms-like tyrosine kinase 1; FRT; Soluble VEGF receptor 1 14; Soluble VEGFR1 variant 2; Soluble VEGFR1 variant 21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7789