Learn More
Biomatik Corporation Human Matrix Metalloproteinase 12 (Mmp-12) Elisa Kit
Supplier:  Biomatik Corporation EKC40963
Encompass_Preferred
Human Matrix Metalloproteinase 12 (Mmp-12) Elisa Kit for detecting Matrix Metallopeptidase 12 (Macrophage Elastase) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P39900; MMP12. Target Synonyms: EC 3.4.24.65; HME; Macrophage elastase; Macrophage metalloelastase; Macrophage metaloelastase; Matrix metallopeptidase 12 (macrophage elastase); Matrix metalloprotease 12; Matrix metalloproteinase-12; ME; MGC138506; MME; MMP 12; MMP-12; Mmp12; MMP12_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7806