Learn More
Biomatik Corporation Rat Podocalyxin,PCX ELISA Kit
Supplier:  Biomatik Corporation EKC41084
Encompass_Preferred
Sample Type: serum, urine, cell culture supernates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9WTQ2. Target Synonyms: Gp200, MGC138240, PC, PCLP. Restriction: For Research Use Only.
Catalog No. 50-228-7821