Learn More
Biomatik Corporation Mouse Interleukin 33(IL-33) ELISA Kit
Supplier:  Biomatik Corporation EKC41149
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 pg/ml - 1000 pg/ml. Sensitivity: 3.9 pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8BVZ5. Target Synonyms: C9orf26, DKFZp586H0523, DVS27, NF-HEV, NFEHEV, RP11-575C20.2, DVS27-related protein|nuclear factor from high endothelial venules. Restriction: For Research Use Only.
Catalog No. 50-228-7827