Learn More
Biomatik Corporation Human Cd70 Antigen (Cd70) Elisa Kit
Supplier:  Biomatik Corporation EKC41426
Encompass_Preferred
Human Cd70 Antigen (Cd70) Elisa Kit for detecting Cd70 Molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P32970; CD70. Target Synonyms: CD 27L; CD 70; CD27 L; CD27 LG; CD27 ligand; CD27-L; CD27L; CD27LG; CD70; CD70 antigen; CD70 molecule; CD70_HUMAN; Ki 24 antigen; Ki24 antigen; Surface antigen CD70; TNFSF 7; TNFSF7; Tumor necrosis factor (ligand) superfamily; member 7; Tumor necrosis factor ligand superfamily member 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7852