Learn More
Biomatik Corporation Mouse Pancreatic Lipase(PL)ELISA Kit
Supplier:  Biomatik Corporation EKC42345
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 mU/mL-4000 mU/mL. Sensitivity: 15.6 mU/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6P8U6. Target Synonyms: PL, pancreatic triacylglycerol lipase|triacylglycerol acylhydrolase. Restriction: For Research Use Only.
Catalog No. 50-228-7874