Learn More
Biomatik Corporation Mouse Alpha-2-macroglobulin(A2M) ELISA kit
Supplier:  Biomatik Corporation EKC42360
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156 μg/mL-10 μg/mL. Sensitivity: 0.039 μg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q61838. Target Synonyms: CPAMD5, DKFZp779B086, FWP007, S863-7,. Restriction: For Research Use Only.
Catalog No. 50-228-7879