Learn More
Biomatik Corporation Human MKI67 (Antigen KI-67) ELISA Kit
Supplier:  Biomatik Corporation EKF57207
Encompass_Preferred
Human MKI67 (Antigen KI-67) ELISA Kit for detecting MKI67 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P46013. Target Synonyms: MKI67; Antigen KI-67; Ki-67; KIA; TSG126; antigen identified by monoclonal Ki-67; KIA; prolifeRation-related Ki-67 antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7888