Learn More
Biomatik Corporation Human ELANE (Neutrophil elastase) ELISA Kit
Supplier:  Biomatik Corporation EKF57218
Encompass_Preferred
Human ELANE (Neutrophil elastase) ELISA Kit for detecting ELANE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08246. Target Synonyms: NE; ELA2; ELANE; Elastase-2; Leukocyte Elastase; Medullasin; Bone marrow serine protease; ELA2granulocyte-derived elastase; elastase 2; neutrophil; elastase; neutrophil expressed; Elastase-2; GE; HLEelastase-2; HNE; Human leukocyte elastase; leukocyte elastase; medullasin; Medullasin; NE; neutrophil elastase; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7890