Learn More
Biomatik Corporation Human CYP3A4 (Cytochrome P450 3A4) ELISA Kit
Supplier:  Biomatik Corporation EKF57229
Encompass_Preferred
Human CYP3A4 (Cytochrome P450 3A4) ELISA Kit for detecting CYP3A4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08684. Target Synonyms: CYP3A4; Cytochrome P450 3A4; CYP3A3 CYPIIIA3; CYPIIIA4; Nifedipine; 8-cineole 2-exo-monooxygenase; Albendazole monooxygenase; Albendazole sulfoxidase; Cytochrome P450 3A3; Cytochrome P450 HLp; Cytochrome P450 NF-25; Cytochrome P450-PCN1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7893