Learn More
Biomatik Corporation Human AMSH (alpha-MSH) ELISA Kit
Supplier:  Biomatik Corporation EKF57232
Encompass_Preferred
Human AMSH (alpha-MSH) ELISA Kit for detecting AMSH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800pg/ml. Sensitivity: 7.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P01189. Target Synonyms: αMSH; Alpha-Melanocyte Stimulating Hormone; AMSH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7895