Learn More
Biomatik Corporation Human VDR (Vitamin D3 receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF57260
Encompass_Preferred
Human VDR (Vitamin D3 receptor) ELISA Kit for detecting VDR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P11473. Target Synonyms: VDR; Vitamin D Receptor; NR1I1; Nuclear receptor subfamily 1 group I member 1; 1; 25-dihydroxyvitamin D3 receptor; vitamin D1; 25- dihydroxyvitamin D3 receptor; vitamin D3 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7901