Learn More
Biomatik Corporation Human SERPINF1 (Pigment epithelium-derived factor) ELISA Kit
Supplier:  Biomatik Corporation EKF57283
Encompass_Preferred
Human SERPINF1 (Pigment epithelium-derived factor) ELISA Kit for detecting SERPINF1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P36955. Target Synonyms: PEDF; Pigment Epithelium Derived Factor; Serpin F1; Pigment epithelium-derived factor; Cell prolifeRation-inducing gene 35 protein; EPC-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7905