Learn More
Biomatik Corporation Human ITGB3 (Integrin beta-3) ELISA Kit
Supplier:  Biomatik Corporation EKF57374
Encompass_Preferred
Human ITGB3 (Integrin beta-3) ELISA Kit for detecting ITGB3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P05106. Target Synonyms: ITGB3; Integrin beta-3; Platelet membrane glycoprotein IIIa; GPIIIa; GP3A; CD61; INGRB3; ITG B3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7930