Learn More
Biomatik Corporation Human GPX3 (Glutathione peroxidase 3) ELISA Kit
Supplier:  Biomatik Corporation EKF57523
Encompass_Preferred
Human GPX3 (Glutathione peroxidase 3) ELISA Kit for detecting GPX3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P22352. Target Synonyms: GPX3; GPX-P; EC 1.11.1; EC 1.11.1.9; Extracellular glutathione peroxidase; glutathione peroxidase 3; glutathione peroxidase 3; plasma; GPx-3; GPXP; GPx-P; GSHPx-3; GSHPx-P; Plasma glutathione peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7957