Learn More
Biomatik Corporation Human PL (Placental Lactogen) ELISA Kit
Supplier:  Biomatik Corporation EKF57689
Encompass_Preferred
Human PL (Placental Lactogen) ELISA Kit for detecting PL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000ng/ml. Sensitivity: 46.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P0DML2. Target Synonyms: PL; Chorionic somatomammotropin hormone; Lactogen; Placental lactogen; Choriomammotropin; CSH1; CSMT; Lactogen; Choriomammotropin; chorionic somatomammotropin hormone; chorionic somatomammotropin hormone 1; placental lactogen; CS-1; CSAchorionic somatomammotropin A; CSH2; CSMT; FLJ75407; hCS-A; Lactogen; lactogen; Placental lactogen; PLchoriomammotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7988