Learn More
Biomatik Corporation Human SMRP (Soluble Mesothelin Related Peptide) ELISA Kit
Supplier:  Biomatik Corporation EKF57716
Encompass_Preferred
Human SMRP (Soluble Mesothelin Related Peptide) ELISA Kit for detecting SMRP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20pmol/ml. Sensitivity: 0.188pmol/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13421. Target Synonyms: SMRP; Soluble Mesothelin Related Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7993