Learn More
Biomatik Corporation Human SPD (Pulmonary Surfactant Associated Protein D) ELISA Kit
Supplier:  Biomatik Corporation EKF57720
Encompass_Preferred
Human SPD (Pulmonary Surfactant Associated Protein D) ELISA Kit for detecting SPD in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35247. Target Synonyms: SPD; Pulmonary Surfactant Associated Protein D; SFTPD; SP-D; Collectin 7; PSP-D; SFTP4; COLEC7; collectin-7; Lung surfactant protein D; PSPD; pulmonary surfactant-associated protein D; pulmonary surfactant apoprotein; surfactant protein D; surfactant; pulmonary-associated protein D; surfactant-associated protein; pulmonary 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7995