Learn More
Biomatik Corporation Human Total P53 (Tumor Protein p53) ELISA Kit
Supplier:  Biomatik Corporation EKF57732
Encompass_Preferred
Human Total P53 (Tumor Protein p53) ELISA Kit for detecting TP53 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04637. Target Synonyms: TP53; BCC7; LFS1; TRP53; Antigen NY-CO-13; FLJ92943; LFS1TRP53; p53; p53 tumor suppressor; P53cellular tumor antigen p53; Phosphoprotein p53; transformation-related protein 53; tumor protein p53; Tumor suppressor p53. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7999