Learn More
Biomatik Corporation Rabbit Procollagen I N-Terminal Propeptide (PINP) ELISA Kit
Supplier:  Biomatik Corporation EKU11356
Encompass_Preferred
Rabbit Procollagen I N-Terminal Propeptide (PINP) ELISA Kit for detecting PINP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 246.9-20,000pg/ml. Sensitivity: 90.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: A0A5F9CPN0. Target Synonyms: P1NP; N-Propeptide Of Type I Procollagen; Procollagen I Amino Terminal Propeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6902