Learn More
Biomatik Corporation Chicken Prolactin (PRL) ELISA Kit
Supplier:  Biomatik Corporation EKU11366
Encompass_Preferred
Chicken Prolactin (PRL) ELISA Kit for detecting PRL in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.92ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P14676. Gene ID: 396453. Target Synonyms: LTH; Luteotropic Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6905