Learn More
Biomatik Corporation Rabbit Nerve Growth Factor (NGF) ELISA Kit
Supplier:  Biomatik Corporation EKU11657
Encompass_Preferred
Rabbit Nerve Growth Factor (NGF) ELISA Kit for detecting NGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G1TXA7. Target Synonyms: NGFB; Beta-NGF; HSAN5; NGF-B; Beta-Nerve Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6979