Learn More
Biomatik Corporation Human Heat Shock Protein 27 (Hsp27) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU11923
Encompass_Preferred
Human Heat Shock Protein 27 (Hsp27) High-sensitive ELISA Kit for detecting Hsp27 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: p04792. Gene ID: 3315. Target Synonyms: HSPB1; HSP-B1; CMT2F; HSP28; Hsp25; Heat Shock 27kDa Protein 1; 28 kDa heat shock protein; Heat shock protein beta-1; Estrogen-regulated 24 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7030