Learn More
Biomatik Corporation Human complement factor H,CFH ELISA Kit
Supplier:  Biomatik Corporation EKC33265
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates.. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2 ng/ml - 2000 ng/ml. Sensitivity: 7.8 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P08603. Target Synonyms: AHUS1, ARMD4, ARMS1, CFHL3, FH, FHL1, HF, HF1, HF2, HUS, MGC88246, H factor 1 (complement)|H factor 2 (complement)|age-related maculopathy susceptibility 1|beta-1-H-globulin|beta-1H|complement facto. Restriction: For Research Use Only.
Catalog No. 50-228-7379