Learn More
Biomatik Corporation Human Creatine Kinase Mm Isoenzyme (Ck-Mm) Elisa Kit
Supplier:  Biomatik Corporation EKC33304
Encompass_Preferred
Human Creatine Kinase Mm Isoenzyme (Ck-Mm) Elisa Kit for detecting Creatine Kinase Mm Isoenzyme(Ck-Mm) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200mU/ml. Sensitivity: 0.78mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7387