Learn More
Biomatik Corporation Human Cyclin-Dependent Kinase Inhibitor 2A, Isoforms 1/2/3 (Cdkn2A) Elisa Kit
Supplier:  Biomatik Corporation EKC33345
Encompass_Preferred
Human Cyclin-Dependent Kinase Inhibitor 2A, Isoforms 1/2/3 (Cdkn2A) Elisa Kit for detecting Cyclin-Dependent Kinase Inhibitor 2A (Melanoma, P16, Inhibits Cdk4) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.69pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P42771; CDKN2A. Target Synonyms: CDKN2A; CDKN2; MTS1; Cyclin-dependent kinase inhibitor 2A; Cyclin-dependent kinase 4 inhibitor A; CDK4I; Multiple tumor suppressor 1; MTS-1; p16-INK4a; p16-INK4; p16INK4A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7392