Learn More
Biomatik Corporation Human E-Selectin Elisa Kit
Supplier:  Biomatik Corporation EKC33632
Encompass_Preferred
Human E-Selectin Elisa Kit for detecting Selectin E in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16581; SELE. Target Synonyms: SELE; ELAM1; E-selectin; CD62 antigen-like family member E; Endothelial leukocyte adhesion molecule 1; ELAM-1; Leukocyte-endothelial cell adhesion molecule 2; LECAM2; CD antigen CD62E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7408